Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30 Mouse anti-Human, FITC, Clone: BERH8, BD

Catalog No. BDB555829 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555829 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555829 Supplier BD Biosciences Supplier No. 555829
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The BerH8 monoclonal antibody specifically binds to CD30. CD30 is a 120 kDa type I transmembrane glycoprotein that is expressed on stimulated T and B cells. CD30 is an activation marker which that initially identified to be expressed on Reed-Sternberg cells. It is also expressed on a few extrafollicular T and B cells located at the rim of germinal centers. CD30 serves as a cytokine receptor. It belongs to the nerve growth factor receptor/tumor necrosis factor receptor (NGFR/TNFR) superfamily and is also known as TNFRSF8. CD30 interaction with CD30 ligand (CD30L/CD153/TNFSF8) can mediate signals for proliferation, apoptosis and cytotoxicity of lymphoid cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD30
Applications Flow Cytometry
Classification Monoclonal
Clone BERH8
Conjugate FITC
Description TNFRSF8; TNR8; CD30L/CD153 receptor; Ki-1; D1S166E
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.