Learn More
CD31 Mouse anti-Human, Alexa Fluor 488, Clone: M89D3, BD
Mouse Monoclonal Antibody
Supplier:  BD Biosciences 558068
Catalog No. BDB558068
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Reacts with CD31, also known as platelet endothelial cell adhesion molecule - 1 (PECAM-1), a sigle chain, type I transmembrane protein of approximately 130 - 140 kDa. CD31 is a member of the immunoglobulin supergene family, its extracellular region is composed of 574 aminoacids and contains six C2-type Ig domains, its intracellular region is composed of 118 residues. CD31 (PECAM-1) is expressed on endothelial cells, platelets, monocytes, neutrophils and NK cells. It has also been observed on subsets of T cells, but not on circulating B cells. It has been implicated in a number of cellular phenomena, including vascular wound healing, angiogenesis, and transendothelial migration of leukocytes and platelet aggregation in inflammatory responses.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only.