Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Mouse anti-Rat, Biotin, Clone: TLD-3A12, BD

Catalog No. BDB555026 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555026 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555026 Supplier BD Biosciences Supplier No. 555026
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TLD-3A12 antibody reacts with CD31, also known as PECAM-1 (platelet endothelial cell adhesion molecule). CD31 is a ∽130 kDa integral membrane glycoprotein, a member of the immunoglobulin super family, that mediates homophilic and heterophilic cell-cell adhesion. CD31 is expressed on endothelial cells, platelets, and subsets of leukocytes. In the human and mouse, multiple alternatively spliced isoforms have been identified, and this alternative splicing may be involved in the regulation of ligand specificity. CD31-mediated endothelial cell-cell interactions are involved in angiogenesis in the mouse and rat. In addition, PECAM-1 has been demonstrated to play an important role in extravasation of leukocytes in vivo, but in vivo treatment with TLD-3A12 mAb had no discernable effect upon the inflammatory infiltrate in experimental allergic encephalomyelitis. The TLD-3A12 mAb partially blocks the proliferation response of antigen-specific CD4+ T cells to antigen-presenting cells and relevant antigen. This mouse anti-rat CD31 antibody cross-reacts with pig endothelial cells and platelets, as determined by immunohistochemical staining of acetone-fixed frozen sections and immunofluorescent staining with flow cytometric analysis, respectively. A suspension of Lewis rat microglial cells were used as the source of the immunogen.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Applications Flow Cytometry
Classification Monoclonal
Clone TLD-3A12
Concentration 0.5mg/mL
Conjugate Biotin
Description PECAM-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Lewis Rat Microglia
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.