Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Mouse anti-Human, PE-Cy7, Clone: WM59, BD

Catalog No. BDB563651 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563651 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563651 Supplier BD Biosciences Supplier No. 563651
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The WM59 monoclonal antibody specifically binds to a 130 kDa glycoprotein, gpIIa', also known as platelet endothelial cell adhesion molecule-1, (PECAM-1). CD31 has wide tissue distribution and is expressed on platelets, monocytes, granulocytes, and in high amounts on endothelial cells. This molecule has been implicated in a number of cellular phenomena, including vascular wound healing and angiogenesis, and transendothelial migration of leukocytes in inflammatory responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Applications Flow Cytometry
Classification Monoclonal
Clone WM59
Conjugate PE-Cyanine7
Description EndoCAM; GPIIA'; PECA1; PECAM1; Platelet endothelial cell adhesion molecule
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.