Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 (PECAM-1) Mouse anti-Human, PE, Clone: L133.1, BD

Catalog No. BDB340297 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340297 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340297 Supplier BD Biosciences Supplier No. 340297
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD31 (Anti-PECAM-1) antibody recognizes the platelet/endothelial cell adhesion molecule-1 (PECAM-1), a 130 to 140-kilodalton (kDa) single-chain integral membrane glycoprotein that is a member of the immunoglobulin gene superfamily. The CD31 antigen is composed of six extracellular immunoglobulin-like domains belonging to the C2 group. C2 domains are also found in other members of the immunoglobulin superfamily, the cell adhesion molecules (CAMs). The CD31 antigen functions as a vascular cell adhesion molecule and is involved in the process of leucocyte migration through the intercellular junctions of vascular endothelial cells. It may also be involved in thrombosis, angiogenesis, wound healing, and inflammation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Applications Flow Cytometry
Classification Monoclonal
Clone L133.1
Concentration 12.5μg/mL
Conjugate PE
Description PECAM-1
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Purified human natural killer cells
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.