Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD314 (NKG2D) Rat anti-Mouse, Brilliant Violet 711, Clone: CX5, BD

Catalog No. BDB563694 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563694 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563694 Supplier BD Biosciences Supplier No. 563694
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The CX5 monoclonal antibody specifically binds to NKG2D, also known as CD314. NKG2D is a lectin-like receptor that is detected on resting and IL-2-activated NK cells, activated CD8-positive T lymphocytes, and LPS-activated macrophages, but not on resting T cells or unstimulated macrophages. NKG2D has little homology to the other members of the NKG2 family, NKG2A, C, and E, and does not form heterodimers with CD94. On NK cells, NKG2D is an activating receptor that associates with DAP10, an adapter protein that stimulates the PI3 kinase pathway. An isoform of mouse NKG2D can also associate with the signaling adapter protein DAP12, also known as KARAP ( K iller cell- A ssociated R eceptor- olypeptide), that activates the Syk and ZAP70 tyrosine kinases. On cytotoxic T cells, NKG2D is a co-stimulatory receptor that associates with DAP10. The ligands for NKG2D include the minor histocompatibility antigen H60, MULT1 ( M urine UL 16-binding protein- L ike T ranscript ), and the five retinoic acid-inducible proteins Rae-1 a , b , g , d and e . Interactions of NKG2D with its ligands are involved in the regulation of innate and immune cytotoxic responses to tumor and pathogen-infected cells and in diabetes progression in the NOD mouse. The CX5 mAb blocks the binding of NKG2D to its ligands.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD314
Applications Flow Cytometry
Classification Monoclonal
Clone CX5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Description Klrk1; NKG2D; NKG2-D; NK cell receptor D; Natural killer cell group 2D
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Purified Mouse NKG2D protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.