Learn More
BioLegend™ anti-CD326, Clone: 9C4
Description
Host Species: Mouse
Clone: 9C4
Isotype: IgG2b, κ
Cross Reactivity: Human
Immunogen: DU.4475 breast carcinoma
Flow Cytometry, Immunofluorescence
Specifications
Specifications
| Antigen | CD326 |
| Classification | Monoclonal |
| Clone | 9C4 |
| Conjugate | FITC |
| Host Species | Mouse |
| Immunogen | DU.4475 breast carcinoma |
| Purification Method | Unpurified |
| Quantity | 100 Tests |
| Primary or Secondary | Primary |
| Target Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.