Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD326 Mouse anti-Human, FITC, Clone: EBA-1, BD

Catalog No. BDB347197 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347197 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347197 Supplier BD Biosciences Supplier No. 347197
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-EpCAM, clone EBA-1, is derived from the fusion of SP2 myeloma cells with spleen cells from BALB/c mice immunized with macromolecular aggregates containing breast carcinoma–associated mucin BCA-225.

Anti-EpCAM is directed against a human epithelial adhesion molecule (EpCAM) that has been identified as a ∽40-kilodalton (kd) protein in Western blotting. This antigen is also known as epithelial surface antigen (ESA) and epithelial glycoprotein 2 (EGP-2).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD326
Applications Flow Cytometry
Classification Monoclonal
Clone EBA-1
Concentration 3μg/mL
Conjugate FITC
Description EpCAM
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Breast carcinoma-associated mucin BCA-225
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 λ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.