Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD328 Mouse anti-Human, Unlabeled, Clone: F023-420, BD

Catalog No. BDB552885 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552885 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552885 Supplier BD Biosciences Supplier No. 552885
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The F023-420 monoclonal antibody specifically recognizes CD328 which is also known as sialic acid binding Ig like lectin 7 (Siglec-7). CD238 is an I-type lectin of approximately 65 kDa, expressed as a monomer on a major subset of NK cells and a subset of CD8+ cells. It is also expressed at moderate levels on monocytes and weakly on granulocytes. Siglecs (sialic acid/immunoglobulin/lectin) are a family of I-type lectins that bind to sialic acids on the cell surface. They are a family of carbohydrate binding proteins within the immunoglobulin superfamily. Siglecs are integral membrane proteins with extracellular N-terminal, V-set Ig domains, followed by variable numbers of C2-set Ig domains.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD328
Applications Flow Cytometry
Classification Monoclonal
Clone F023-420
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Siglec-7
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Siglec-7
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.