Learn More
CD328 Mouse anti-Human, PE, Clone: F023-420, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 558372
Description
The F023-420 monoclonal antibody specifically recognizes CD328 which is also known as sialic acid binding Ig like lectin 7 (Siglec-7). CD238 is an I-type lectin of approximately 65 kDa, expressed as a monomer on a major subset of NK cells and a subset of CD8+ cells. It is also expressed at moderate levels on monocytes and weakly on granulocytes. Siglecs (sialic acid/immunoglobulin/lectin) are a family of I-type lectins that bind to sialic acids on the cell surface. They are a family of carbohydrate binding proteins within the immunoglobulin superfamily. Siglecs are integral membrane proteins with extracellular N-terminal, V-set Ig domains, followed by variable numbers of C2-set Ig domains.

Specifications
CD328 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Recombinant Siglec-7 | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
F023-420 | |
Siglec-7 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.