Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD329 (Siglec-9) Mouse anti-Human, PE, Clone: E10-286, BD

Catalog No. BDB564251 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564251 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564251 Supplier BD Biosciences Supplier No. 564251
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The E10-286 monoclonal antibody specifically binds to CD329. CD329 is also known as Sialic acid-binding Ig-like lectin 9 (Siglec-9), formerly known as Siglec-Y, which is expressed on peripheral blood monocytes, granulocytes and NK cells. Siglecs (sialic acid/immunolgobulin/lectin) are a family of I-type lectins binding to sialic acids on the cell surface. They are a family of carbohydrate-binding proteins within the immunoglobulin superfamily. Siglecs are all integral membrane proteins with extracellular domains consisting of unusual N-termial V-set Ig domains, followed by variable numbers of C2-set Ig domains.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD329
Applications Flow Cytometry
Classification Monoclonal
Clone E10-286
Conjugate PE
Description Siglec-9; Siglec Y; FOAP-9; OBBP-LIKE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.