Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD33 Mouse anti-Human, PerCP-Cy5.5, Clone: P67.6, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 341650
Description
CD33, clone P67.6, is derived from the hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with FMY9S5 cells containing the CD33 gene.
CD33 recognizes a human myelomonocytic antigen, Mr 67 kilodaltons (kd).
Specifications
| CD33 | |
| Monoclonal | |
| 6.3μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| FMY9S5 cells containing the CD33 gene | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| P67.6 | |
| PerCP-Cyanine5.5 | |
| Mouse | |
| 50 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction