Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD335 (NKp46) Mouse anti-Human, Brilliant Violet 711, Clone: 9E2/Nkp46, BD

Catalog No. BDB563043 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563043 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563043 Supplier BD Biosciences Supplier No. 563043
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 9E2/Nkp46 monoclonal antibody specifically binds to CD335. CD335 is also known as the Natural killer cell p46-related protein (NKp46) and the Natural cytotoxicity triggering receptor 1 (NCR1). CD335 is a 46 kDa type I membrane glycoprotein that is expressed on resting and activated NK cells. Its extracellular region contains two C2-type, Ig-like domains. The transmembrane domain contains a positively charged amino acid (Arg) which could be involved in stabilizing the association with CD3ζ. Its intracellular region does not contain immunoreceptor tyrosine-based activating motifs (ITAM), but it is linked to intracytoplasmic transduction machinery by its association with CD3ζ and FcεRIγadaptor proteins. CD335 along with NKp30 and NKp44 are referred to as natural cytotoxicity receptors (NCR). These receptors play very important roles in cells that kill virus-infected target cells, tumor cells and MHC-class I-unprotected cells.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD335
Applications Flow Cytometry
Classification Monoclonal
Clone 9E2/Nkp46
Conjugate Brilliant Violet 711
Description NCR1; NK-p46; hNKp46; LY94; Natural cytotoxicity triggering receptor 1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NKp46 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.