Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD337 (NKp30) Mouse anti-Human, PE, Clone: P30-15, BD

Catalog No. BDB558407 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558407 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558407 Supplier BD Biosciences Supplier No. 558407
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody clone p30-15 reacts with CD337, also known as NKp30, a receptor found on the surface of natural killer (NK) cells. NK cells are large lymphoid cells discovered because of their ability to recognize and kill abnormal cells such as tumor and virally infected cells. NK cell immune responses are regulated by a balance of activating and inhibitory signals generated by cell surface receptors. Inhibitory receptors recognize MHC class I molecules on normal cells producing a negative signal to the NK cell. Loss of MHC class I expression in infected or transformed cells results in the loss of this negative signal leading to NK cell activation. In concert with the loss of inhibitory signals, activation signals via NK receptors such as NKp30, NKp44, NKp46, NKG2D, and NKp80 mediate the activation of NK cells. NKp30 cooperates with NKp46 and/or NKp44 in the induction of NK cell-mediated cytotoxicity against the majority of target cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD337
Applications Flow Cytometry
Classification Monoclonal
Clone P30-15
Concentration 0.2mg/mL
Conjugate PE
Description NCR3; Natural cytotoxicity triggering receptor 3; NKp30; MALS; LY117; 1C7
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NKp30 e tracellular domain Transfected Cell Line
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.