Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD338 Mouse anti-Human, APC, Clone: 5D3, BD

Catalog No. BDB561451 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561451 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561451 Supplier BD Biosciences Supplier No. 561451
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 5D3 monoclonal antibody specifically binds to an epitope of ABCG2 (BCRP1), a multi-drug resistance protein that is a member of ATP binding cassette (ABC) transporters. It is highly expressed on primitive stem cells as identified by the “side-population” (SP) phenotype. This SP phenotype is based on the efflux of fluorescent dyes such as Rhodamine 123 and Hoechst 33342. The expression of ABCG2 appears to be highly conserved as it has been identified in various species. Studies show that highly purified murine stem cells express BCRP1 mRNA and this expression declines sharply as the stem cells express CD34. The highest levels of BCRP1 mRNA expression have been seen in KDR+ human stem cells. ABCG2/BCRP1 was clustered as CD338 in the HLDA8 workshop.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD338
Applications Flow Cytometry
Classification Monoclonal
Clone 5D3
Conjugate APC
Description ABCG2; ABCP; BCRP; BMDP; MXR1; ABC15; BCRP1; CDw338; EST157481; MGC102821
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ABCG2-transfected Mouse cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.