Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CD34 Class II, Clone: QBEND/10, ™ (Chemicon™)
Supplier: MilliporeSigma™ CBL496F/DEL
Description
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation

Specifications
CD34 Class II | |
FITC | |
Human endothelial vesicles | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 Assays | |
Inflammation, Immunology | |
Human, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction