Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Class III Mouse anti-Human, Clone: 581, MilliporeSigma™

Catalog No. CBL555MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL555MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL555MI Supplier MilliporeSigma Supplier No. CBL555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-CD34 Antibody, Class III, clone 581 is validated for use in FC for the detection of CD34 Class III.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD34 Class III
Applications Flow Cytometry
Classification Monoclonal
Clone 581
Conjugate Unconjugated
Formulation Protein G affinity chromatography. The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working t
Gene Accession No. NM_001025109.1; NM_001773.2
Gene Symbols CD34
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.