Learn More
CD34 Mouse anti-Human, PE-CF594, Clone: 563, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 562449
Description
The 563 monoclonal antibody specifically binds to CD34. CD34 is a single-chain 105-120 kDa heavily O-glycosylated transmembrane glycoprotein expressed on hematopoietic progenitor cells, vascular endothelium and some tissue fibroblasts. The intracellular chain of the CD34 antigen is a target for phosphorylation by activated protein kinase C suggesting that CD34 may play a role in signal transduction. CD34 may play a role in adhesion of specific antigens to endothelium. Clone 563 belongs to the class III epitope; it is resistant to neuraminidase, chymopapain and glycoprotease. This antibody is able to block the binding of other CD34 mAbs like 581.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).
Specifications
| CD34 | |
| Monoclonal | |
| PE-CF594 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Cynomolgus, Rhesus Monkey | |
| IgG1 κ |
| Flow Cytometry | |
| 563 | |
| gp 105-120 | |
| Mouse | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.