Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 (gp 105-120) Mouse anti-Human, Unlabeled, Clone: MY10, BD

Catalog No. BDB347660 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347660 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347660 Supplier BD Biosciences Supplier No. 347660
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD34 (Anti–HPCA-1), clone My10, is derived from the hybridization of mouse Sp2/0-Ag14 cells with spleen cells of BALB/c mice immunized with the human cell line KG-1a.

The CD34 antigen is a single-chain transmembrane glycoprotein, Mr 105 to 120 kilodaltons (kd). The antigen is associated with human hematopoietic progenitor cells and is a differentiation stage–specific leucocyte antigen. Clone My10 and clone 8G12 recognize two distinct CD34 epitopes; at least three epitopes have been identified.

The CD34 antigen is a single-chain transmembrane glycoprotein, Mr 105 to 120 kilodaltons (kd). The antigen is associated with human hematopoietic progenitor cells and is a differentiation stage–specific leucocyte antigen. Clone My10 and clone 8G12 recognize two distinct CD34 epitopes; at least three epitopes have been identified.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD34 (gp 105-120)
Applications Flow Cytometry
Classification Monoclonal
Clone MY10
Concentration 50μg/mL
Conjugate Unconjugated
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human cell line KG-1a
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.