Learn More
CD34 Mouse anti-Human, PE, Clone: 8G12, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 348057
Description
CD34 (Anti–HPCA-1), clone My10, is derived from the hybridization of mouse Sp2/0-Ag14 cells with spleen cells of BALB/c mice immunized with the human cell line KG-1a.
The CD34 antigen is a single-chain transmembrane glycoprotein, Mr 105 to 120 kilodaltons (kd). The antigen is associated with human hematopoietic progenitor cells and is a differentiation stage–specific leucocyte antigen. Clone My10 and clone 8G12 recognize two distinct CD34 epitopes; at least three epitopes have been identified.
The CD34 antigen is a single-chain transmembrane glycoprotein, Mr 105 to 120 kilodaltons (kd). The antigen is associated with human hematopoietic progenitor cells and is a differentiation stage–specific leucocyte antigen. Clone My10 and clone 8G12 recognize two distinct CD34 epitopes; at least three epitopes have been identified.
Specifications
| CD34 | |
| Monoclonal | |
| 25μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human cell line KG-1a | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. | |
| IgG1 κ |
| Flow Cytometry | |
| 8G12 | |
| PE | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.