Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Mouse anti-Human, PE, Clone: 563, BD

Catalog No. BDB550619 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550619 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550619 Supplier BD Biosciences Supplier No. 550619
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone 563 reacts with the human form of the 105-120 kDa heavily O-glycosylated and N-glycosylated transmembrane glycoprotein, CD34, expressed on stem/progenitor hematopoietic cells, endothelial cells and some tissue. Clone 563 also cross reacts with a subset of peripheral blood mononuclear cells of both rhesus and cynomolgus macaque monkeys, but not baboon. The distribution of stem-like cells is similar to that observed with peripheral blood mononuclear cells from normal human donors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD34
Applications Flow Cytometry
Classification Monoclonal
Clone 563
Conjugate PE
Description gp 105-120
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.