Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD344 Mouse anti-Human, Unlabeled, Clone: CH3A4A7, BD

Catalog No. BDB563657 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563657 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563657 Supplier BD Biosciences Supplier No. 563657

Mouse Monoclonal Antibody

The CH3A4A7 monoclonal antibody reacts with an extracellular epitope on human CD344 which is also known as Frizzled family receptor 4 (FZD4) or Frizzled-4. CD344 is a member of the Frizzled family of proteins within the G protein-coupled receptor (GPCR) superfamily. CD344 is a receptor for Wnt proteins and participates in the activation of the Wnt/beta-catenin pathway (canonical Wnt signaling). Patients with familial exudative vitreoretinopathy (FEVR) carry mutations in Frizzled-4. This disease is characterized by retinal degeneration and hearing loss. On tissues, CD344 is almost ubiquitously expressed. CD344 is highly expressed in adult heart and skeletal muscle.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD344
Applications Flow Cytometry
Classification Monoclonal
Clone CH3A4A7
Conjugate Unconjugated
Description FZD4; Frizzled family receptor 4; Frizzled-4; Fz-4; FzE4; hFz4; EVR1; FEVR
Host Species Mouse
Immunogen Human neuroblastoma Cell Line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.