Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Rat anti-Mouse, Biotin, Clone: 8C12, BD

Catalog No. BDB553816 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553816 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553816 Supplier BD Biosciences Supplier No. 553816
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 8C12 antibody recognizes an epitope present on the 190-kDa complementreceptor protein, originally designated CR1 (CD35), but not the 145-150-kDa CR2 (CD21) molecule. Unlike the human system, in which these proteins are products of independent genes, both of these mouse receptors are membraneproteins resulting from the alternative splicing of mRNA transcribed from the Cr2 gene. Therefore, an alternative nomenclature has been proposed, designating the proteins Cr2-190 (CD21b) and Cr2-145 (CD21a), respectively. The epitope recognized by 8C12 mAb is only present on CD35/CD21b.Moreover, it has also been proposed that Crry is the true mouse genetic homologue of human CR1 (CD35). In the mouse, CD35 is expressed on the majority of peripheral B cells, on the majority of resident peritoneal macrophages, on peripheral blood granulocytes after treatment with N-formyl-Met-Leu-Phe, and on follicular dendritic cells, but not on thymocytes, T cells, erythrocytes, or platelets. In addition, it has not been detected, at the protein or mRNA level, in the macrophage cell line J774, bone marrow-derived macrophages, or thioglycollate-elicited peritoneal macrophages. The 8C12 mAb has been reported to inhibit rosette formation by C3bbearing sheep erythrocytes,to block the complement-dependent trapping of immune complexes by follicular dendritic cells,and to down-regulate mouse CD35 expression upon in vivo application,inhibiting only some primary antibody responses to immunization. B lymphocytes of Cr2[

*Please note that the isotype of 8C12 mAb was originally reported to be Rat IgG2c. Further investigations have demonstrated that the isotype of 8C12 mAb is Rat IgG2a.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD35
Applications Flow Cytometry
Classification Monoclonal
Clone 8C12
Concentration 0.5mg/mL
Conjugate Biotin
Description CR1, CD21b
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Purified mouse CR1
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.