Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Mouse anti-Human, FITC, Clone: E11, BD

Catalog No. BDB555452 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555452 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555452 Supplier BD Biosciences Supplier No. 555452
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with CR1, the receptor for the complement component C3b, composed of four different allotypes (160, 190, 220 and 250 kD). CD35 antigen is expressed on granulocytes, monocytes, B cells, some NK cells and erythrocytes. It binds complement components C3b and C4b, mediating phagocytosis by neutrophils and monocytes. This antibody cannot inhibit the phagocytic capacity of granulocytes. CD35 antibody is useful in studies of complement receptors.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD35
Applications Flow Cytometry
Classification Monoclonal
Clone E11
Conjugate FITC
Description CR1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Cells of the Monocyte Lineage
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.