Learn More
Description
Reacts with CR1, the receptor for the complement component C3b, composed of four different allotypes (160, 190, 220 and 250 kD). CD35 antigen is expressed on granulocytes, monocytes, B cells, some NK cells and erythrocytes. It binds complement components C3b and C4b, mediating phagocytosis by neutrophils and monocytes. This antibody cannot inhibit the phagocytic capacity of granulocytes. CD35 antibody is useful in studies of complement receptors.
Specifications
Specifications
| Antigen | CD35 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | E11 |
| Conjugate | PE |
| Description | CR1 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human Cells of the Monocyte Lineage |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.