Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD354 (TREM-1) Mouse anti-Human, Alexa Fluor 647, Clone: 193015, BD

Catalog No. BDB564472 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564472 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564472 Supplier BD Biosciences Supplier No. 564472
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 193015 monoclonal antibody specifically binds to CD354 which is also known as TREM-1 (Triggering Receptor Expressed on Myeloid Cells 1 or Triggering Receptor Expressed on Monocytes 1). CD354 is a type I transmembrane glycoprotein that belongs to the immunoglobulin superfamily. CD354 is expressed by neutrophils and a subset of monocytes. Its expression is upregulated on leucocytes in response to microbial lipopolysaccharide, eg, during microbial infections or sepsis. CD354 activation triggers the release of proinflammatory cytokines and chemokines.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD354
Applications Flow Cytometry
Classification Monoclonal
Clone 193015
Conjugate Alexa Fluor 647
Description TREM1, Triggering receptor expressed on myeloid cells 1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human TREM-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.