Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36 Mouse anti-Human, APC, Clone: CB38 (NL07), BD

Catalog No. BDB550956 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550956 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550956 Supplier BD Biosciences Supplier No. 550956
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CB38 monoclonal antibody specifically binds to CD36. CD36 is a 88 kDa glycoprotein IV (GPIV), the receptor for extracellular matrix proteins such as collagen and thrombospondin. CD36 is known to mediate the adhesion of Plasmodium falciparum. CD36 antigen is expressed on monocytes, platelets, endothelial cells, and some human tumor cell lines but not on lymphocytes and granulocytes. It is a very early marker of erythroid differentiation. CD36 antibody induces degranulation, release of ATP and serotonin, increase in [Ca2+]ι, and tyrosine phosphorylation of a substrate protein of 130 kDa.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD36
Applications Flow Cytometry
Classification Monoclonal
Clone CB38 (NL07)
Conjugate APC
Description GPIIIb; Platelet GPIV; OKM5-antigen; PASIV
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Platelet Extracts
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.