Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36 Mouse anti-Mouse, BB515, Clone: CRF D-2712, BD

Catalog No. BDB565094 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565094 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565094 Supplier BD Biosciences Supplier No. 565094
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CRF D-2712 monoclonal antibody specifically binds to mouse and rat CD36, an 88-kDa glycoprotein member of the class B scavenger receptor family. CD36 is a multifunctional receptor that has a variety of ligands and is expressed on many cell types. Consequently, it is known by several names, including macrophage scavenger receptor, platelet glycoprotein IV, and fatty acid translocase. Cells that express CD36 include platelets, megakaryocytes, monocytes/macrophages, dendritic cells, erythroid precursors, microvascular endothelia, skeletal, cardiac, and smooth muscle cells, adipocytes, and epithelia of retina, breast, and intestine. This multi-ligand receptor functions in the clearance of apoptotic cells and abnormal erythrocytes (in malaria and sickle-cell anemia), the transport of long-chain fatty acids and oxidized low-density lipoproteins, the regulation of angiogenesis by mediating the effects of thrombospondin-1, and the mediation of coronary vasoconstriction. Crosslinking of cell-surface CD36 with plate-bound CRF D-2712 antibody promotes internalization of photoreceptor outer segment fragments by cultured retinal pigment epithelium.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD36
Applications Flow Cytometry
Classification Monoclonal
Clone CRF D-2712
Concentration 0.2mg/mL
Conjugate Brilliant Blue 515 (BB515)
Description Platelet glycoprotein 4; PAS-4; GPIIIB; Fatty acid translocase; Scavenger R
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Recombinant mouse CD36
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgA κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.