Learn More
CD37 Mouse anti-Human, Alexa Fluor 647, Clone: M-B371, BD
Catalog No. BDB561562 Shop All BD Biosciences Products
Change view
Click to view available options
50 Tests; Alexa Fluor 647
Catalog No.
BDB561562 50 Tests; Alexa Fluor 647
1 options
This item is not returnable. View return policy
Catalog No. BDB561562 Supplier BD Biosciences Supplier No. 561562
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mouse Monoclonal Antibody
The M-B371 monoclonal antibody reacts with a 40-52 kDa type II integral membrane glycoprotein present on B cells from the pre-B-cell stage but not on plasma cells. CD37 is also expressed on activated, proliferating cells of germinal centers. There is low expression of CD37 on some T and myeloid cells. The function of CD37 has not yet been clearly identified.
TRUSTED_SUSTAINABILITY
WARNING: Reproductive Harm - www.P65Warnings.ca.gov