Learn More
CD37 Mouse anti-Human, PerCP-Cy5.5, Clone: M-B371, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 561563
Description
The M-B371 monoclonal antibody reacts with a 40-52 kDa type II integral membrane glycoprotein present on B cells from the pre-B-cell stage but not on plasma cells. CD37 is also expressed on activated, proliferating cells of germinal centers. There is low expression of CD37 on some T and myeloid cells. The function of CD37 has not yet been clearly identified.

Specifications
CD37 | |
Monoclonal | |
PerCP-Cyanine5.5 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
M-B371 | |
GP52-40; Tetraspanin-26; TSPAN26; Tspan-26 | |
Mouse | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.