Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD37 Mouse anti-Human, V450, Clone: M-B371, BD

Catalog No. BDB561548 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561548 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561548 Supplier BD Biosciences Supplier No. 561548

Mouse Monoclonal Antibody

The M-B371 monoclonal antibody reacts with a 40-52 kDa type II integral membrane glycoprotein present on B cells from the pre-B-cell stage but not on plasma cells. CD37 is also expressed on activated, proliferating cells of germinal centers. There is low expression of CD37 on some T and myeloid cells. The function of CD37 has not yet been clearly identified.

The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450 nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD37
Applications Flow Cytometry
Classification Monoclonal
Clone M-B371
Conjugate Violet Fluor 450 (V450)
Description GP52-40; Tetraspanin-26; TSPAN26; Tspan-26
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.