Learn More
CD38 Rat anti-Mouse, Biotin, Clone: 90, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 553762
Description
The 90 monoclonal antibody specifically binds to CD38, a 42 kDa transmembrane glycoprotein on immature and mature, resting and activated, B lymphocytes. In contrast to humans, CD38 expression is down-regulated on mouse germinal center B cells and plasma cells. CD38 is also expressed on a subpopulation of thymic and peripheral T cells, NK cells, and splenic macrophages. Furthermore, CD38 has been detected on bone marrow-derived hematopoietic stem cells. The CD38 molecule is reported to exhibit both cyclase and hydrolase activities and plays a role in lymphocyte activation. CD31, both human and mouse, is reported to be a ligand for human CD38.

Specifications
CD38 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Bone Marrow Pre-B cells | |
0.5 mg | |
Immunology | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
90 | |
Biotin | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.