Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Mouse anti-Human, Unlabeled, Clone: 22, BD

Catalog No. BDB611114 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611114 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611114 Supplier BD Biosciences Supplier No. 611114
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lymphocyte trafficking is essential for efficient immune responses in that it allows antigen sensitive lymphocytes to encounter antigen and be recruited to sites of infection. Trafficking involves the passage of lymphocytes across the HEV, a process that is mediated by selectins. CD38 is a single-chain type II transmembrane glycoprotein that is expressed by a variety of immature and activated lymphocytes. It mediates a selectin-like binding to endothelial cells, thus functioning as an adhesion molecule. Human and mouse forms of the protein are highly similar and homologous to the Aplysia californica ADP-ribosyl-cyclase. CD38 catalyzes the synthesis of cyclic ADP-ribose (cADPR), a putative endogenous regulator of calcium release. This is thought to be key to CD38-mediated signal transduction. A possible ligand for CD38 is the Ig superfamily protein CD31. In addition, activation of CD38 with a monoclonal antibody induces leukemic cell proliferation. Thus, CD38 is a cell surface protein that is important for lymphocyte migration and may be involved in the propagation of leukemic cells in AML (acute myelogenous leukemia).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD38
Applications Western Blot
Classification Monoclonal
Clone 22
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD38 aa. 171-292
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.