Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioLegend™ anti-CD38, Clone: 90

Catalog No. NC9905804
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC9905804 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC9905804 Supplier BioLegend™ Supplier No. 102718
Add to Cart
Add to Cart

Host Species: Rat
Clone: 90.0
Isotype: IgG2a, κ
Cross Reactivity: Mouse
Immunogen: Mouse bone marrow pre-B cells

Flow Cytometry

Specifications

Antigen CD38
Classification Monoclonal
Clone 90.0
Concentration 0.2mg/mL
Conjugate PE-Cyanine7
Host Species Rat
Immunogen Mouse bone marrow pre-B cells
Purification Method Unpurified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.