Learn More
Description
Host Species: Mouse
Clone HIT2
Isotype IgG1, κ
Species Reactivity Hu
Flow Cytometry
Specifications
Specifications
| Antigen | CD38 |
| Classification | Monoclonal |
| Clone | HIT2 |
| Conjugate | Brilliant Violet 510 |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.