Learn More
CD38, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA532323
Description
CD38 (cluster of differentiation 38), also known as cyclic ADP ribose hydrolase, is a transmembrane glycoprotein found on the surface of many immune cells including plasma cells, activated or immature T and B cells, monocytes, and natural killer cells. CD38 participates in cell adhesion, signal transduction and calcium signaling. It is expressed at high levels in the pancreas, liver, kidney, malignant lymphoma and neuroblastoma. CD38 is used as one of the plasma cell markers and its ligand is CD31 molecules.
Heat-mediated antigen retrieval is recommended prior to staining, using an EDTA buffer for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is tonsil tissue.
Specifications
CD38 | |
Polyclonal | |
CD38 | |
T10, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, CD38 antigen p45, NAD+ nucleosidase, cADPr hydrolase 1, cyclic ADP-ribose hydrolase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
952 | |
Store at 4°C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
P28907 | |
CD38 | |
Synthetic peptide near the C-terminus of human CD38. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.