Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD39 Mouse anti-Mouse, Clone: 5F2, MilliporeSigma™

Catalog No. 04973MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-973-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-973-MI Supplier MilliporeSigma Supplier No. 04973
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-CD39 Antibody, clone 5F2 is validated for use in flow cytometry, western blotting for the detection of CD39.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD39
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5F2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_033978
Gene Symbols CD39, Ecto-apyrase, Ectoapyrase
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse CD39.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.