Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD39 Mouse anti-Human, PE, Clone: TÜ66, BD

Catalog No. BDB555464 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555464 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555464 Supplier BD Biosciences Supplier No. 555464
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TU66 monoclonal antibody reacts with human CD39 also known as ectonucleoside triphosphate diphosphohydrolase 1 (ENTPD1) an ectoenzyme that degrades ATP to AMP. It is a member of the family of ectonucleoside triphosphate dihydrolases (E-NTPDases) known to be involved in regulation of extracellular nucleotide catabolism, controlling the extracellular nucleoside triphosphate pool (NTP). CD39 is expressed on a subset of T cells, B cells and dendritic cells with weak staining of monocytes and granulocytes. Recently, CD39 has been found to be expressed primarly by immune-suppressive Foxp3(+) regulatory T (Treg) cells in both human and mice. In humans, CD39 is restricted to a subset of Foxp3+ regulatory effector/memory-like T cells. In mice, the enzyme is present on most if not all CD4+CD25+ cells and CD39 expression is driven by Foxp3. It is thought that CD39 allows Treg cells to enter inflamed areas where high levels of ATP are present.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD39
Applications Flow Cytometry
Classification Monoclonal
Clone TÜ66
Conjugate PE
Description ENTPD1; NTPDase-1; Ecto-ATPase 1; Ecto-ATPDase 1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.