Learn More
CD3d molecule, delta (CD3-TCR complex), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CD3D protein.
Supplier: Abnova Corporation H00000915B01
Description
The protein encoded by this gene is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The encoded membrane protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in this gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms have been found for this gene. Other variants may also exist, but the full-length natures of their transcripts has yet to be defined. [provided by RefSeq
Sequence: MEHSTFLSGLVLATLLSQVSPFKIPIEELEDRVFVNCNTSITWVEGTVGTLLSDITRLDLGKRILDPRGIYRCNGTDIYKDKESTVQVHYRMCQSCVELDPATVAGIIVTDVIATLLLALGVFCFAGHETGRLSGAADTQALLRNDQVYQPLRDRDDAQYSHLGGNWARNKSpecifications
CD3d | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human CD3D protein. | |
CD3D | |
CD3-DELTA/T3D | |
Mouse | |
50 μL | |
Membrane Receptors | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_000732 | |
CD3D | |
CD3D (NP_000723, 1 a.a. ∼ 171 a.a) full-length human protein. | |
RUO | |
Yes | |
915 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.