Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3e Hamster anti-Mouse, Unlabeled, Clone: 500A2, BD

Catalog No. BDB550277 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550277 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550277 Supplier BD Biosciences Supplier No. 550277
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The 500A2 monoclonal antibody specifically binds to the 25-kDa ε chain of the T-cell receptor-associated CD3 complex expressed on mouse thymocytes, mature T lymphocytes, and NKT cells. Plate-bound and soluble forms of the 500A2 antibody can activate T cells in vitro . Activation of a mouse T-cell clone by the 500A2 antibody can be blocked by Fab fragments of the GK1.5 anti-CD4 antibody. This suggests that the 500A2 antibody may bind an epitope on CD3e close to a site at which CD4 associates with the T-cell receptor. The 500A2 antibody reportedly does not to crossreact with rat leukocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3e
Applications Flow Cytometry
Classification Monoclonal
Clone 500A2
Concentration 62.5μg/mL
Conjugate Unconjugated
Description CD3; CD3 epsilon; Cd3e; CD3ε; T3e
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse T-cell receptor
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.