Learn More
CD3e Hamster anti-Mouse, Pacific Blue, Clone: 500A2, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 558214
Description
This antibody has been reported to react with the 25-kDa ε chain of the T-cell receptor-associated CD3 complex expressed on thymocytes, mature T lymphocytes, and NK-T cells on many mouse stains. Plate-bound and soluble hamster anti-mouse CD3e (clone 500A2) antibody has been reported to activate unprimed T cells in vitro. Activation of a mouse T-cell clone by soluble hamster anti-mouse CD3e (clone 500A2) can be blocked by Fab fragments of anti-CD4 antibody GK1.5, suggesting that the hamster anti-mouse CD3e (clone 500A2) antibody may bind an epitope of CD3e close to a site at which CD4 associates with the T-cell receptor. This hamster mAb to a mouse leukocyte antigen has been reported not to cross-react with rat leukocytes.

Specifications
CD3e | |
Monoclonal | |
0.2mg/mL | |
CD3; CD3 epsilon; Cd3e; CD3ε; T3e | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
500A2 | |
Pacific Blue | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Mouse T-cell receptor | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.