Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3e Mouse anti-Pig, PE-Cy7, Clone: BB23-8E6-8C8, BD

Catalog No. BDB561477 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561477 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561477 Supplier BD Biosciences Supplier No. 561477
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The BB23-8E6-8C8 monoclonal antibody specifically binds to the 25-kDa ε chain of the T-cell receptor-associated CD3 complex. It recognizes all CD4+ and most CD8+ peripheral blood T lymphocytes, most thymocytes and phytohemagglutinin-stimulated blasts, and subsets of spleen and Peyer’s patch lymphocytes. BB23-8E6-8C8 is a immunoglobulin isotype switch variant of the BB23-8E6 clone. This isotype-switch variant induces a proliferative response of peripheral blood mononuclear cells. The epitope recognized by BB23-8E6 mAb was designated CD3a by the Second International Swine CD Workshop.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3e
Applications Flow Cytometry
Classification Monoclonal
Clone BB23-8E6-8C8
Concentration 0.2mg/mL
Conjugate PE-Cyanine7
Description CD3 epsilon subunit; CD3e; T-cell surface glycoprotein CD3 epsilon chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Pig peripheral blood mononuclear cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.