Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3e Syrian Hamster anti-Mouse, V500, Clone: 500A2, BD

Catalog No. BDB560771 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560771 0.1 mg
BDB560773 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560771 Supplier BD Biosciences Supplier No. 560771
Add to Cart
Add to Cart

Syrian Hamster Monoclonal Antibody

This antibody has been reported to react with the 25kDa ε chain of the T-cell receptor-associated CD3 complex expressed on thymocytes, mature T lymphocytes, and NK-T cells on many mouse stains. Plate-bound and soluble hamster anti-mouse CD3e (clone 500A2) antibody has been reported to activate unprimed T cells in vitro. Activation of a mouse T-cell clone by soluble hamster anti-mouse CD3e (clone 500A2) can be blocked by Fab fragments of anti-CD4 antibody GK1.5, suggesting that the hamster anti-mouse CD3e (clone 500A2) antibody may bind an epitope of CD3e close to a site at which CD4 associates with the T-cell receptor. This hamster mAb to a mouse leukocyte antigen has been reported not to cross-react with rat leukocytes. The antibody is conjugated to BD Horizon™ V500, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser with an Ex max of 415 nm and Em Max at 500 nm. BD Horizon V500 conjugates emit at a similar wavelength to Amcyan yet exhibit reduced spillover into the FITC channel. For more information on BD Horizon V500, visit bdbiosciences.com/colors. When compensating dyes in this spectral range (such as Horizon™ V500 and AmCyan), the most accurate compensation can be obtained using single stained cellular controls. Due to spectral differences between cells and beads in this channel, using BD CompBeads can result in spillover errors for V500 and AmCyan reagents. Therefore, the use of BD CompBeads or BD CompBeads Plus to determine spillover values for these reagents is not recommended. Different V500 reagents (e.g. CD4 vs. CD45) can have slightly different fluorescence spillover therefore, it may also be necessary to use clone specific compensation controls when using these reagents.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3e
Applications Flow Cytometry
Classification Monoclonal
Clone 500A2
Concentration 0.2mg/mL
Conjugate Violet Fluor 500 (V500)
Description CD3ε chain
Formulation Aqueous buffered solution containing protein stabilizer, glycerol and ≤0.09% sodium azide.
Host Species Syrian Hamster
Immunogen Mouse T-cell receptor
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.