Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD4 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
$116.00 - $221.50
Specifications
Antigen | CD4 |
---|---|
Clone | 11830 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB3791A025
![]() |
R&D Systems
FAB3791A025 |
25 Tests |
Each for $116.00
|
|
|||||
FAB3791A100
![]() |
R&D Systems
FAB3791A100 |
100 Tests |
Each for $221.50
|
|
|||||
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry.Specifications
CD4 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
CD4 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CD4 in direct ELISAs. |
11830 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
P01730 | |
920 | |
Recombinant Human CD4. Extracellular Domain Accession # P01730 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title