Learn More
CD4 Mouse anti-Human, APC-H7, Clone: RPA-T4, BD
Mouse Monoclonal Antibody
$170.70 - $366.00
Specifications
Antigen | CD4 |
---|---|
Clone | RPA-T4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC-H7 |
Description
The RPA-T4 clone reacts with CD4, a 59kDa single-chain transmembrane glycoprotein (receptor for human immunodeficiency virus (HIV)) present on T-helper/inducer cell populations. This antibody binds to the D1 domain (CDR1 and CDR3 epitopes) of the CD4 antigen and reacts with approximately 80% of thymocytes and 45% of peripheral blood lymphocytes. CD4 is also present in low density on peripheral blood monocytes. RPA-T4 is capable of blocking HIV-1, gp120, and inhibits syncytium formation.
Host Species: Mouse
Clone: RPA-T4
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
CD4 | |
Flow Cytometry | |
APC-H7 | |
RUO | |
Human | |
IgG1 κ | |
Affinity Purified |
RPA-T4 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.