Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human, Brilliant Violet 786, Clone: L200, BD

Catalog No. BDB563914 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563914 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563914 Supplier BD Biosciences Supplier No. 563914
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The L200 monoclonal antibody specifically binds to the human form of the 56 kDa transmembrane glycoprotein, CD4, present on the T-helper/inducer subset of normal human donor peripheral blood lymphocytes. The L200 antibody also crossreacts with a subset of CD3-positive peripheral blood lymphocytes, but not monocytes, of both Rhesus and Cynomolgus Macaque monkeys. Crossreactivity on both lymphocytes and monocytes (weak) from Baboons is also observed. The distribution on lymphocytes is similar for both human and monkey cells, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon™ BV786 can be excited by the violet laser and detected in a filter used to detect Cy7™-like dyes (eg, 780/60-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone L200
Conjugate Brilliant Violet 786
Description L3T4 ; T-cell surface antigen T4/Leu-3; W3/25 ; CD4 antigen (p55)
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human HPB-ALL Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.