Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD4, BUV395; Clone: GK1.5; BD
Supplier: BD Biosciences 563790
Description
Applications:
Flow cytometry

Specifications
CD4 | |
Monoclonal | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
Primary | |
Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze |
Flow Cytometry | |
GK1.5 | |
Brilliant Ultraviolet 395 | |
CTL clone V4 | |
RUO | |
Mouse | |
IgG2b κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction