Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human, Clone: 024-10D6.B3, MilliporeSigma™

Catalog No. MAB3706MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3706MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3706MI Supplier MilliporeSigma Supplier No. MAB3706
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect CD4 using this Anti-CD4 Antibody, intracellular, clone 024-10D6.B3 validated for use in ELISA, EMSA, FUNC and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Electromobility Shift Assay, ELISA, Functional Assay, Immunocytochemistry
Classification Monoclonal
Clone 024-10D6.B3
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02Mphosphate buffer, 0.25M NaCl, with 0.1% sodium azide.
Gene Accession No. NM_000616.3
Gene Symbols CD4, CD4mut
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 439-458 of the cytoplasmic tail of human CD4.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.