Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Feline, Lion, Biotin, Clone: 3-4F4, Southern Biotech™
SDP

Catalog No. OB813008 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB8130-08 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB8130-08 Supplier Southern Biotechnology Associates Supplier No. 813008
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SouthernBiotech offers anti-feline primary antibodies which are suitable flow cytometry and microscopy applications.

Specifications

Antigen CD4
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 3-4F4
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Supplied in PBS/NaN3
Host Species Mouse
Immunogen Feline T cells
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Feline, Lion
Content And Storage Store at 2-8°C
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.