Learn More
BioLegend™ anti-CD4, Clone: GK1.5
Supplier: BioLegend™ 100434

Description
Host Species: Rat
Clone: GK1.5
Isotype: IgG2b, κ
Cross Reactivity: Mouse
Immunogen: Mouse CTL clone V4
Flow Cytometry
Specifications
CD4 | |
GK1.5 | |
PerCP-Cyanine5.5 | |
Mouse CTL clone V4 | |
100 μg | |
Mouse |
Monoclonal | |
0.2mg/mL | |
Rat | |
Unpurified | |
Primary | |
IgG2b κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.